Guanidinium carbonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Guanidine carbonate salt, Diguanidinium carbonate, Guanidine carbonate

A12853-0B A12853-30 A12853-0I
AAA12853-0BEA 85 USD
AAA12853-0B AAA12853-30 AAA12853-0I
Guanidinium carbonate 98%
Guanidinium carbonate

Purity: 98% CAS No.: 593-85-1 Molecular Formula: C3H12N6O3 Formula Weight: 180.17 Melting Point: ca 198° dec. Beilstein Reference: 3628359

Formula: C₃H₁₂N₆O₃
MW: 180.17 g/mol
Melting Pt: ∼ 198 °C (dec.)
Density: 1.251 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013029
CAS Number: 593-85-1
EINECS: 209-813-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR