D-(+)-Galactose 98%

Supplier: Thermo Scientific Chemicals

Synonyms: D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal

A12813-30 A12813-0B A12813-18
AAA12813-30EA 132.48 USD
AAA12813-30 AAA12813-0B AAA12813-18
D-(+)-Galactose 98%
D-(+)-Galactose

Purity: 98% CAS No.: 59-23-4 Molecular Formula: C6H12O6 Formula Weight: 180.16 Melting Point: 164-168° Beilstein Reference: 1724619 Merck Reference: 14,4335 Optical Rotation: +80° (c=5 in water, 24h) MDL No.: MFCD00151230

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 527 °C (1013 hPa)
Melting Pt: 164…168 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151230
CAS Number: 59-23-4
EINECS: 200-416-4
Merck Index: 13,04356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&vid=3c02003b1d12c9bfd11c2e6faf5cc37d4325d208&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR