4-Isopropoxybenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: ALDH1A3-IN-3

A12760-06 A12760-03 A12760-14
AAA12760-06EA 98.8 USD
AAA12760-06 AAA12760-03 AAA12760-14
4-Isopropoxybenzaldehyde 97%
4-Isopropoxybenzaldehyde

Purity: 97% CAS No.: 18962-05-5 Molecular Formula: C10H12O2 Formula Weight: 164.21 Refractive Index: 1.5470 Boiling Point: 108-110°/5mm MDL No.: MFCD00052357

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 108…110 °C/5 mm
Storage Temperature: Ambient
MDL Number: MFCD00052357
CAS Number: 18962-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR