α,α,α-Trifluoro-3,4-diaminotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Trifluoromethyl)-1,2-phenylenediamine, 4-(Trifluoromethyl)benzene-1,2-diamine, 3,4-Diaminobenzotrifluoride, 4-(Trifluoromethyl)-o-phenylenediamine
A12749-03
A12749-06
AAA12749-14EA
655.58
USD
AAA12749-14
AAA12749-03
AAA12749-06
α,α,α-Trifluoro-3,4-diaminotoluene 98%
α,α,α-Trifluoro-3,4-diaminotoluene
Purity: 98%
CAS No.: 368-71-8
Molecular Formula: C7H7F3N2
Formula Weight: 176.14
Melting Point: 58-62°
Beilstein Reference: 909258
UN No.: 2811
MDL No.: MFCD00042456
Formula:
C₇H₇F₃N₂ MW: 176.14 g/mol Boiling Pt: 101 - 102 °C (7 -rr) Melting Pt: 58…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042456 CAS Number: 368-71-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...