2-Butylthiophene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12740-14
A12740-06
AAA12740-14EA
201
USD
AAA12740-14
AAA12740-06
AAA12740-22
2-Butylthiophene 98+%
2-Butylthiophene
Purity: 98+%
CAS No.: 1455-20-5
Molecular Formula: C8H12S
Formula Weight: 140.25
Flash Point: 56°(133°F)
Refractive Index: 1.5020
Odor: Recognizable
Boiling Point: 181-182°
Beilstein Reference: 107046
UN No.: 1993
MDL No.: MFCD00022534
Formula:
C₈H₁₂S MW: 140.25 g/mol Boiling Pt: 181…182 °C Density: 0.957 g/cm³ (19 °C) Flash Pt: 56 °C (133 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00022534 CAS Number: 1455-20-5 EINECS: 215-935-1 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...