4-Hydroxybenzhydrazide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxybenzoic hydrazide, 4-Hydroxybenzohydrazide, p-Hydroxybenzoic acid hydrazide, 4-Hydroxybenzene-1-carbohydrazide, 4-Hydroxybenzoic acid hydrazide

A12702-22 A12702-14 A12702-36
AAA12702-22EA 118 USD
AAA12702-22 AAA12702-14 AAA12702-36
4-Hydroxybenzhydrazide 98%
4-Hydroxybenzhydrazide

Purity: 98% CAS No.: 5351-23-5 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Melting Point: ca 267° dec. Beilstein Reference: 473052

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 370 °C (1013 hPa)
Melting Pt: ∼ 267 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007605
CAS Number: 5351-23-5
EINECS: 226-326-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR