2-Bromo-3-hydroxypyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

A12687-06 A12687-22 A12687-14
AAA12687-06EA 58.8 USD
AAA12687-06 AAA12687-22 AAA12687-14
2-Bromo-3-hydroxypyridine 99%
2-Bromo-3-hydroxypyridine

Purity: 99% CAS No.: 6602-32-0 Molecular Formula: C5H4BrNO Formula Weight: 174.00 Melting Point: 182-186° Beilstein Reference: 109829

Formula: C₅H₄BrNO
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 185 - 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006220
CAS Number: 6602-32-0
EINECS: 229-547-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR