2',3'-Dichloroacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2,3-Dichlorophenyl)ethanone

A12667-03 A12667-06
AAA12667-03EA 39.4 USD
AAA12667-03 AAA12667-14 AAA12667-06
2',3'-Dichloroacetophenone 98%
2',3'-Dichloroacetophenone

Purity: 98% CAS No.: 56041-57-7 Molecular Formula: C8H6Cl2O Formula Weight: 189.04 Refractive Index: 1.5600 Boiling Point: 106-108°/2mm MDL No.: MFCD00052988

Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 106…108 °C/2 mm
Density: 1.293
Storage Temperature: Ambient
MDL Number: MFCD00052988
CAS Number: 56041-57-7
EINECS: 259-954-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR