5-Fluoro-2-methylbenzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12641-03
A12641-06
AAA12641-14EA
539.92
USD
AAA12641-14
AAA12641-03
AAA12641-06
5-Fluoro-2-methylbenzenesulfonyl chloride 97%
5-Fluoro-2-methylbenzenesulphonyl chloride
Purity: 97%
CAS No.: 445-05-6
Molecular Formula: C7H6ClFO2S
Formula Weight: 208.64
Flash Point:
Formula:
C₇H₆ClFO₂S Boiling Pt: 130…132 °C/15 mm Density: 1.43 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00052958 CAS Number: 445-05-6 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...