Phthalimide 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Phthalic acid imide
A12584-0E
A12584-0C
AAA12584-0EEA
101
USD
AAA12584-0E
AAA12584-0C
Phthalimide 99%
Phthalimide
MDL: MFCD00005881
Beilstein: 118522
Slightly soluble in water. Soluble in aqueous alkali hydroxides
Purity: 99%
CAS No.: 85-41-6
Molecular Formula: C8H5NO2
Formula Weight: 147.14
Flash Point: 165°(329°F)
Odor: Odorless
pH: 3.8
Melting Point: 232-236°
Boiling Point: 366°
Solubility: Slightly soluble in water. Soluble in aqueous alkali hydroxides
Beilstein Reference: 118522
Merck Reference: 14,7373
MDL No.: MFCD00005881
Formula:
C₈H₅NO₂ MW: 147.13 g/mol Boiling Pt: 366 °C Melting Pt: 232…236 °C Density: 1.21 g/cm³ (20 °C) Flash Pt: 165 °C(329 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005881 CAS Number: 85-41-6 EINECS: 201-603-3 Merck Index: 12,07529 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...