Dimethyl succinate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
DBE-4 dibasic ester, Succinic acid dimethyl ester, 1,4-Butanedioic acid dimethyl ester
A12565-0B
A12565-22
A12565-30
AAA12565-0BEA
250
USD
AAA12565-0B
AAAA12565-22
AAA12565-30
Dimethyl succinate 98+%
Dimethyl succinate
Purity: 98+%
CAS No.: 106-65-0
Molecular Formula: C6H10O4
Formula Weight: 146.14
Flash Point: 85°(185°F)
Refractive Index: 1.4190
Melting Point: 18-19°
Boiling Point: 196°
Beilstein Reference: 956776
Merck Reference: 14,8869
MDL No.: MFCD00008466
Formula:
C₆H₁₀O₄ MW: 146.14 g/mol Boiling Pt: 200 Melting Pt: 18…19 °C Density: 1.119 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008466 CAS Number: 106-65-0 EINECS: 203-419-9 Merck Index: 14,8869 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...