3'-Hydroxyacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

A12547-36 A12547-22
AAA12547-36EA 267 USD
AAA12547-36 AAA12547-22
3'-Hydroxyacetophenone 98%
3'-Hydroxyacetophenone

Purity: 98% CAS No.: 121-71-1 Molecular Formula: C8H8O2 Formula Weight: 136.15 Melting Point: 93-97° Boiling Point: 296° MDL No.: MFCD00002298

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 86 - 88 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002298
CAS Number: 121-71-1
EINECS: 204-494-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR