Glycolic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12511-36
A12511-22
AAA12511-36EA
298.7
USD
AAA12511-36
AAA12511-22
Glycolic acid 98%
Glycolic acid
Purity: 98%
CAS No.: 79-14-1
Molecular Formula: C2H4O3
Formula Weight: 76.05
Physical Form: Crystalline powder
Density: 1.49
Appearance: White
Odor: Unpleasant
pH: 2
Melting Point: 74-80°
Beilstein Reference: 1209322
Merck Reference: 14,4498
UN No.: 3261
MDL No.: MFCD00004312
Formula:
C₂H₄O₃ Boiling Pt: 266 °C (1013 hPa) Melting Pt: 79 - 80 °C Density: 1.49 g/cm³ (20 °C) Flash Pt: ≥ 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004312 CAS Number: 79-14-1 EINECS: 201-180-5 UN: 3261 ADR: 8,II Merck Index: 13,04511 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...