Cyclobutanol 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12430-14
A12430-03
A12430-06
AAAA12430-14EA
1112
USD
AAAA12430-14
AAA12430-03
AAA12430-06
Cyclobutanol 96%
Cyclobutanol
Purity: 96%
CAS No.: 2919-23-5
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: 21°(69°F)
Refractive Index: 1.4360
Boiling Point: 123-124°
Beilstein Reference: 2035937
UN No.: 1120
MDL No.: MFCD00001318
Formula:
C₄H₈O Boiling Pt: 123…124 °C Density: 0.925 Flash Pt: 21 °C (69 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001318 CAS Number: 2919-23-5 EINECS: 220-858-1 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...