2,4,6-Tribromophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

A12424-30 A12424-0B
AAA12424-30EA 46.9 USD
AAA12424-30 AAA12424-0B
2,4,6-Tribromophenol 98%
2,4,6-Tribromophenol

Purity: 98% CAS No.: 118-79-6 Molecular Formula: C6H3Br3O Formula Weight: 330.80 Melting Point: 86-92° Boiling Point: 282-290° subl. Merck Reference: 14,9613 UN No.: 3077 MDL No.: MFCD00002150

Formula: C₆H₃Br₃O
MW: 330.8 g/mol
Boiling Pt: 282…290 °C (subl.)
Melting Pt: 86…92 °C
Density: 2.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002150
CAS Number: 118-79-6
EINECS: 204-278-6
UN: 3077
ADR: 9,III
Merck Index: 12,09744

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR