Cyclopentanecarboxylic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12375-18
A12375-30
A12375-09
AAA12375-18EA
150
USD
AAA12375-18
AAA12375-30
AAA12375-09
Cyclopentanecarboxylic acid 99%
Cyclopentanecarboxylic acid
Purity: 99%
CAS No.: 3400-45-1
Molecular Formula: C6H10O2
Formula Weight: 114.14
Flash Point: 93°(199°F)
Refractive Index: 1.4540
Melting Point: -7°
Boiling Point: 215-216°
Beilstein Reference: 2039729
MDL No.: MFCD00001371
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 215…216 °C Melting Pt: –7 °C Density: 1.05 g/cm³ (20 °C) Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001371 CAS Number: 3400-45-1 EINECS: 222-269-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...