3-Methylpyrocatechol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Methylcatechol, 3-Methylbenzene-1,2-diol
A12324-14
A12324-22
AAA12324-14EA
123
USD
AAA12324-14
AAA12324-22
3-Methylpyrocatechol 98%
3-Methylpyrocatechol
Purity: 98%
CAS No.: 488-17-5
Molecular Formula: C7H8O2
Formula Weight: 124.14
Flash Point: 140°(284°F)
Melting Point: 62-68°
Boiling Point: 241°
Beilstein Reference: 774602
MDL No.: MFCD00016435
Formula:
C₇H₈O₂ MW: 124.14 g/mol Boiling Pt: 241 °C Melting Pt: 62…68 °C Flash Pt: 140 °C(284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00016435 CAS Number: 488-17-5 EINECS: 207-672-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...