o-Cresol 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Hydroxy-2-methylbenzene, 2-Methylphenol, 2-Hydroxytoluene

A12307-36 A12307-0P
AAA12307-36EA 53.8 USD
AAA12307-36 AAA12307-0P
o-Cresol 98+%
o-Cresol

Purity: 98+% CAS No.: 95-48-7 Molecular Formula: C7H8O Formula Weight: 108.14 Flash Point: 81°(177°F) Odor: Phenol-like Boiling Point: 190-192° Beilstein Reference: 506917 Merck Reference: 14,2579 UN No.: 3455 MDL No.: MFCD00002226

Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 190…192 °C
Melting Pt: 30…34 °C
Density: 1.047 g/cm³ (20 °C)
Flash Pt: 81 °C(177 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002226
CAS Number: 95-48-7
EINECS: 202-423-8
UN: 3455
ADR: "6.1",II
Merck Index: 13,02604

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR