1-Bromo-2,4-dichlorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4-Dichlorobromobenzene

A12298-06 A12298-22 A12298-14
AAA12298-06EA 44.4 USD
AAA12298-06 AAA12298-22 AAA12298-14
1-Bromo-2,4-dichlorobenzene 98%
1-Bromo-2,4-dichlorobenzene

Purity: 98% CAS No.: 1193-72-2 Molecular Formula: C6H3BrCl2 Formula Weight: 225.91 Melting Point: 26-27° Boiling Point: 235° MDL No.: MFCD00018333

Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 235 °C
Melting Pt: 26…27 °C
Storage Temperature: Ambient
MDL Number: MFCD00018333
CAS Number: 1193-72-2
EINECS: 214-778-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR