2-Bromo-5-methoxybenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12261-18
A12261-30
A12261-09
AAA12261-18EA
250
USD
AAA12261-18
AAAA12261-30
AAA12261-09
2-Bromo-5-methoxybenzoic acid 98+%
2-Bromo-5-methoxybenzoic acid
Purity: 98+%
CAS No.: 22921-68-2
Molecular Formula: C8H7BrO3
Formula Weight: 231.05
Odor: Odorless
Melting Point: 156-160°
Beilstein Reference: 639763
MDL No.: MFCD00020214
Formula:
C₈H₇BrO₃ Melting Pt: 156…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020214 CAS Number: 22921-68-2 EINECS: 245-329-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...