3,4-Dichlorophenol 99%

Supplier: Thermo Scientific Chemicals
Warning

A12256-14 A12256-22
AAA12256-14EA 76.7 USD
AAA12256-14 AAA12256-22
3,4-Dichlorophenol 99%
3,4-Dichlorophenol

Purity: 99% CAS No.: 95-77-2 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Odor: Phenol-like Melting Point: 64-68° Boiling Point: 252-255° Beilstein Reference: 1907693 UN No.: 2020 MDL No.: MFCD00002258

Formula: C₆H₄Cl₂O
Boiling Pt: 252…255 °C
Melting Pt: 64…68 °C
Density: 1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002258
CAS Number: 95-77-2
EINECS: 202-450-5
UN: 2020
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR