2,3-Dichloropyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

A12241-14 A12241-06
AAA12241-14EA 168 USD
AAA12241-14 AAA12241-22 AAA12241-06
2,3-Dichloropyridine 99%
2,3-Dichloropyridine

Purity: 99% CAS No.: 2402-77-9 Molecular Formula: C5H3Cl2N Formula Weight: 147.99 Odor: Unpleasant Melting Point: 65-69° Beilstein Reference: 109811 UN No.: 2811 MDL No.: MFCD00006229

Formula: C₅H₃Cl₂N
Boiling Pt: 202.5 - 203.5 °C (1013 hPa)
Melting Pt: 65…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00006229
CAS Number: 2402-77-9
EINECS: 219-281-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR