(2-Aminoethoxy)benzene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
ω-Aminophenetole, 2-Phenoxyethylamine, 2-Aminoethyl phenyl ether, β-Aminophenetole
A12225-14
A12225-06
AAAA12225-14EA
446
USD
AAAA12225-14
AAA12225-06
(2-Aminoethoxy)benzene 98%
(2-Aminoethoxy)benzene
Organic intermediate
Purity: 98%
CAS No.: 1758-46-9
Molecular Formula: C8H11NO
Formula Weight: 137.18
Flash Point: 107°(224°F)
Refractive Index: 1.5370
Odor: Amine-like
Boiling Point: 88-90°/1mm
Beilstein Reference: 774671
UN No.: 2735
MDL No.: MFCD00052975
Formula:
C₆H₅OCH₂CH₂NH₂ MW: 137.18 g/mol Boiling Pt: 88…90 °C/1 mm Density: 1.048 g/cm³ (20 °C) Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00052975 CAS Number: 1758-46-9 EINECS: 217-153-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...