4-Nitropyrocatechol 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Nitrocatechol, 4-Nitrobenzene-1,2-diol

A12122-22 A12122-06 A12122-14
AAAA12122-22EA 830 USD
AAAA12122-22 AAA12122-06 AAA12122-14
4-Nitropyrocatechol 98+%
4-Nitropyrocatechol

MDL: MFCD00007242 Beilstein Registry No.: 1867508

Formula: C₆H₅NO₄
MW: 155.11 g/mol
Melting Pt: 174…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00007242
CAS Number: 3316-09-4
EINECS: 222-009-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR