4-Acetoxy-3-methoxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Formyl-2-methoxyphenyl acetate, Vanillin acetate

A12103-30 A12103-22 A12103-14
AAAA12103-30EA 283 USD
AAAA12103-30 AAA12103-22 AAA12103-14
4-Acetoxy-3-methoxybenzaldehyde 98%
4-Acetoxy-3-methoxybenzaldehyde

MDL: MFCD00003362 Beilstein Registry No.: 1963795

Formula: C₁₀H₁₀O₄
Melting Pt: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00003362
CAS Number: 881-68-5
EINECS: 212-920-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR