2-Benzoylthiophene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12036-18
A12036-09
A12036-06
AAA12036-18EA
172
USD
AAA12036-18
AAA12036-09
AAAA12036-06
2-Benzoylthiophene 98%
2-Benzoylthiophene
MDL: MFCD00022495
Beilstein Registry No.: 3929
Purity: 98%
CAS No.: 135-00-2
Molecular Formula: C11H8OS
Formula Weight: 188.25
Melting Point: 55-57°
Boiling Point: 124-126°/0.5mm
MDL No.: MFCD00022495
Formula:
C₁₁H₈OS Boiling Pt: 124…126 °C/0.5 mm Melting Pt: 55…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022495 CAS Number: 135-00-2 EINECS: 205-169-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...