Dimethoxy(dimethyl)silane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dimethyldimethoxysilane
A12025-22
A12025-14
AAA12025-22EA
93.4
USD
AAA12025-22
AAA12025-14
Dimethoxy(dimethyl)silane 97%
Dimethoxy(dimethyl)silane
MDL: MFCD00025691
Beilstein Registry No.: 1733694
Purity: 97%
CAS No.: 1112-39-6
Molecular Formula: C4H12O2Si
Formula Weight: 120.22
Flash Point: -8°(17°F)
Refractive Index: 1.3710
Melting Point: -80°
Boiling Point: 81-82°
Beilstein Reference: 1733694
UN No.: 1993
MDL No.: MFCD00025691
Formula:
C₄H₁₂O₂Si MW: 120.22 g/mol Boiling Pt: 82 Melting Pt: -80 Density: 0.867 Flash Pt: –8 °C (17 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025691 CAS Number: 1112-39-6 EINECS: 214-189-4 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...