Pentafluoroiodobenzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Iodopentafluorobenzene
A12000-06
A12000-18
A12000-09
AAAA12000-06EA
34.7
USD
AAAA12000-06
AAA12000-18
AAA12000-09
Pentafluoroiodobenzene 97%
Pentafluoroiodobenzene
MDL: MFCD00001032
Beilstein Registry No.: 2051549
Purity: 97%
CAS No.: 827-15-6
Molecular Formula: C6F5I
Formula Weight: 293.96
Refractive Index: 1.4960
Melting Point: -29°
Boiling Point: 161-163°
Beilstein Reference: 2051549
MDL No.: MFCD00001032
Formula:
C₆F₅I MW: 293.96 g/mol Boiling Pt: 161…163 °C Melting Pt: –29 °C Density: 2.2055 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001032 CAS Number: 827-15-6 EINECS: 212-565-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...