N,N-Dimethyl-p-toluidine 99%

Supplier: Thermo Scientific Chemicals
Danger

A11988-22 A11988-36 A11988-0E
AAA11988-22EA 40.7 USD
AAA11988-22 AAA11988-36 AAAA11988-0E
N,N-Dimethyl-p-toluidine 99%
N,N-Dimethyl-p-toluidine

MDL: MFCD00008316 Beilstein Registry No.: 774409


Purity: 99% CAS No.: 99-97-8 Molecular Formula: C9H13N Formula Weight: 135.21 Flash Point: 83°(181°F) Refractive Index: 1.5460 Melting Point: -25° Boiling Point: 210-211° Beilstein Reference: 774409 UN No.: 1708 MDL No.: MFCD00008316

Formula: C₉H₁₃N
Boiling Pt: 210…211 °C
Melting Pt: –25 °C
Density: 0.937
Flash Pt: 83 °C (181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008316
CAS Number: 99-97-8
EINECS: 202-805-4
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR