2'-Bromoacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Bromoacetophenone

A11982-09 A11982-18 A11982-30
AAA11982-09EA 76 USD
AAA11982-09 AAA11982-18 AAA11982-30
2'-Bromoacetophenone 99%
2'-Bromoacetophenone

MDL: MFCD00000067 Beilstein Registry No.: 1931534


Purity: 99% CAS No.: 2142-69-0 Molecular Formula: C8H7BrO Formula Weight: 199.05 Flash Point: 110°(230°F) Refractive Index: 1.5680 Boiling Point: 116-117°/12mm Beilstein Reference: 1931534 MDL No.: MFCD00000067

Formula: C₈H₇BrO
MW: 199.05 g/mol
Boiling Pt: 116…117 °C/12 mm
Melting Pt: 0 °C
Density: 1.476 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000067
CAS Number: 2142-69-0
EINECS: 218-398-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR