4'-Hydroxyacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

A11974-36 A11974-22 A11974-0B
AAAA11974-36EA 170 USD
AAAA11974-36 AAAA11974-22 AAA11974-0B
4'-Hydroxyacetophenone 99%
4'-Hydroxyacetophenone

MDL: MFCD00002359 Beilstein Registry No.: 774355


Purity: 99% CAS No.: 99-93-4 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 166°(330°F) Melting Point: 106-108° Boiling Point: 286° Beilstein Reference: 774355 MDL No.: MFCD00002359

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 286 °C
Melting Pt: 106…108 °C
Density: 1.139 g/cm³ (25 °C)
Flash Pt: 166 °C(330 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002359
CAS Number: 99-93-4
EINECS: 202-802-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR