2,4-Dimethylimidazole 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11949-14
A11949-06
AAA11949-14EA
375
USD
AAA11949-14
AAA11949-06
2,4-Dimethylimidazole 97%
2,4-Dimethylimidazole
MDL: MFCD00022365
Beilstein Registry No.: 1639
Purity: 97%
CAS No.: 930-62-1
Molecular Formula: C5H8N2
Formula Weight: 96.13
Melting Point: 84-88°
Boiling Point: 266°/733mm
UN No.: 3263
MDL No.: MFCD00022365
Formula:
C₅H₈N₂ MW: 96.13 g/mol Boiling Pt: 266 °C (733 -rr) Melting Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022365 CAS Number: 930-62-1 EINECS: 213-221-4 UN: 3263 ADR: 8,III |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...