Cyclopropanecarbonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cyclopropanecarboxylic chloride, Cyclopropanecarboxylic acid chloride
A11948-14
A11948-22
AAA11948-14EA
44.5
USD
AAA11948-14
AAA11948-22
Cyclopropanecarbonyl chloride 98%
Cyclopropanecarbonyl chloride
MDL: MFCD00001277
Beilstein Registry No.: 471286
Purity: 98%
CAS No.: 4023-34-1
Molecular Formula: C4H5ClO
Formula Weight: 104.54
Flash Point: 23°(73°F)
Refractive Index: 1.4520
Odor: Pungent
Boiling Point: 119-120°
Beilstein Reference: 471286
UN No.: 2920
MDL No.: MFCD00001277
Formula:
C₄H₅ClO MW: 104.54 g/mol Boiling Pt: 119…120 °C Density: 1.15 g/cm³ (20 °C) Flash Pt: 23 °C (73 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001277 CAS Number: 4023-34-1 EINECS: 223-684-4 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...