1,3-Propanesultone 99%

Supplier: Thermo Scientific Chemicals
Danger

A11923-09 A11923-18 A11923-30
AAA11923-09EA 44.2 USD
AAA11923-09 AAA11923-18 AAA11923-30
1,3-Propanesultone 99%
1,3-Propanesultone

MDL: MFCD00005355 Beilstein Registry No.: 109782 Notes: Special handling precautions required. View MSDS prior to purchase.


Purity: 99% CAS No.: 1120-71-4 Molecular Formula: C3H6O3S Formula Weight: 122.14 Flash Point: 110°(230°F) Melting Point: 30-33° Boiling Point: 179-181°/30mm Beilstein Reference: 109782 UN No.: 2811 MDL No.: MFCD00005355

Formula: C₃H₆O₃S
Boiling Pt: 179…181 °C/30 mm
Melting Pt: 30…33 °C
Density: 1.392
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005355
CAS Number: 1120-71-4
EINECS: 214-317-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR