trans-4-Hydroxy-L(-)-proline 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH

A11851-22 A11851-14 A11851-09
AAA11851-22EA 229 USD
AAA11851-22 AAA11851-14 AAAA11851-09
trans-4-Hydroxy-L(-)-proline 99+%
trans-4-Hydroxy-L(-)-proline

MDL: MFCD00064320 Beilstein Registry No.: 471933 Optical Rotation: -76°C (c=4 in water)


Purity: 99+% CAS No.: 51-35-4 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: 270-273° dec. Beilstein Reference: 471933 Merck Reference: 14,4840 Optical Rotation: -76° (c=4 in water) MDL No.: MFCD00064320

Formula: C₅H₉NO₃
MW: 131.13 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 270…273 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064320
CAS Number: 51-35-4
EINECS: 200-091-9
Merck Index: 13,04866

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR