2-Acetyl-5-chlorothiophene 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-(5-Chloro-2-thienyl)-1-ethanone, Rivaroxaban Impurity 20
A11850-09
A11850-18
AAA11850-09EA
38.4
USD
AAA11850-09
AAA11850-18
AAA11850-30
2-Acetyl-5-chlorothiophene 99%
2-Acetyl-5-chlorothiophene
MDL: MFCD00005444
Beilstein Registry No.: 113936
Purity: 99%
CAS No.: 6310-09-4
Molecular Formula: C6H5ClOS
Formula Weight: 160.62
Flash Point: 108°(226°F)
Melting Point: 44-48°
Boiling Point: 117-118°/17mm
Beilstein Reference: 113936
UN No.: 2811
MDL No.: MFCD00005444
Formula:
C₆H₅ClOS MW: 160.62 g/mol Boiling Pt: 117…118 °C/17 mm Melting Pt: 44…48 °C Flash Pt: 108 °C(226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005444 CAS Number: 6310-09-4 EINECS: 228-630-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...