Nitromethane 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00007400
Beilstein Registry No.: 1698205
Fieser: 1,739 4,357 9,323 12,337 13,199
Purity: 98+%
CAS No.: 75-52-5
Molecular Formula: CH3NO2
Formula Weight: 61.04
Flash Point: 35°(95°F)
Physical Form: Liquid
Density: 1.130
Refractive Index: 1.3820
Appearance: Colorless
Odor: Characteristic
Melting Point: -29°
Boiling Point: 101°
Beilstein Reference: 1698205
Merck Reference: 14,6611
UN No.: 1261
MDL No.: MFCD00007400
Formula:
CH₃NO₂ Boiling Pt: 101 °C Melting Pt: –29 °C Density: 1.138 g/cm³ (20 °C) Flash Pt: 35 °C (95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007400 CAS Number: 75-52-5 EINECS: 200-876-6 UN: 1261 ADR: 3,II Merck Index: 13,06644 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...