2-Bromoaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11788-22
A11788-36
A11788-14
AAA11788-22EA
177
USD
AAA11788-22
AAAA11788-36
AAA11788-14
2-Bromoaniline 98%
2-Bromoaniline
MDL: MFCD00007632
Purity: 98%
CAS No.: 615-36-1
Molecular Formula: C6H6BrN
Formula Weight: 172.03
Flash Point:
Formula:
C₆H₆BrN MW: 172.02 g/mol Boiling Pt: 228…230 °C Melting Pt: 27…31 °C Density: 1.86 g/cm³ (20 °C) Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007632 CAS Number: 615-36-1 EINECS: 210-421-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...