Methyl-4-aminobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-aminobenzoate

A11715-14 A11715-22 A11715-36
AAA11715-14EA 70.1 USD
AAA11715-14 AAA11715-22 AAA11715-36
Methyl-4-aminobenzoate 98%
Methyl-4-aminobenzoate

MDL: MFCD00007891 Beilstein Registry No.: 775913


Purity: 98% CAS No.: 619-45-4 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 110-114° Beilstein Reference: 775913

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 110…114 °C
Density: 1.216 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007891
CAS Number: 619-45-4
EINECS: 210-598-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR