N-Bromophthalimide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11601-06 A11601-22 A11601-14
AAA11601-06EA 33.09 USD
AAA11601-06 AAA11601-22 AAA11601-14
N-Bromophthalimide 98+%
N-Bromophthalimide

MDL: MFCD00005888 Beilstein Registry No.: 131211


Purity: 98+% CAS No.: 2439-85-2 Molecular Formula: C8H4BrNO2 Formula Weight: 226.03 Melting Point: 204-208° Beilstein Reference: 131211

Formula: C₈H₄BrNO₂
Melting Pt: 204…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00005888
CAS Number: 2439-85-2
EINECS: 219-467-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR