1,2,4-Triazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Useful as an antifungal compound
Purity: 99%
CAS No.: 288-88-0
Molecular Formula: C2H3N3
Formula Weight: 69.07
Flash Point: 158°(316°F)
Melting Point: 118-122°
Boiling Point: 260°
Beilstein Reference: 104767
Merck Reference: 14,9605
MDL No.: MFCD00005228
Formula:
C₂H₃N₃ Boiling Pt: 260 °C Melting Pt: 118…122 °C Density: 1.394 g/cm³ (25 °C) Flash Pt: 158 °C(316 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005228 CAS Number: 288-88-0 EINECS: 206-022-9 Merck Index: 12,09735 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...