Bromocyclohexane 98%
Supplier: Thermo Scientific Chemicals
Warning
|
MDL: MFCD00003819
Beilstein Registry No.: 1098373
Purity: 98%
CAS No.: 108-85-0
Molecular Formula: C6H11Br
Formula Weight: 163.06
Flash Point: 62°(143°F)
Refractive Index: 1.4950
Melting Point: -57°
Boiling Point: 166-167°
Beilstein Reference: 1098373
Merck Reference: 14,2730
MDL No.: MFCD00003819
Formula:
C₆H₁₁Br Boiling Pt: 166…167 °C Melting Pt: –57 °C Density: 1.335 Flash Pt: 62 °C(143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003819 CAS Number: 108-85-0 EINECS: 203-622-2 Merck Index: 14,2730 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...