4-Chlorobenzamidine hydroiodide 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chlorobenzene-1-carboximidamide hydroiodide, 4-Chlorophenylamidine hydroiodide

A11571-06 A11571-03
AAA11571-06EA 127 USD
AAA11571-06 AAA11571-03 AAAA11571-14
4-Chlorobenzamidine hydroiodide 96%
4-Chlorobenzamidine hydroiodide

MDL: MFCD00051987 Beilstein Registry No.: 8280383


Purity: 96% CAS No.: 115297-57-9 Molecular Formula: C7H7ClN2.HI Formula Weight: 282.51 Melting Point: ca 235° dec. Beilstein Reference: 8280383

Formula: C₇H₈ClIN₂
MW: 282.51 g/mol
Melting Pt: ca 235 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051987
CAS Number: 115297-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR