3-Bromobenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

A11507-03 A11507-14 A11507-06
AAA11507-03EA 66.7 USD
AAA11507-03 AAAA11507-14 AAA11507-06
3-Bromobenzenesulfonyl chloride 97%
3-Bromobenzenesulphonyl chloride

MDL: MFCD00052313 Beilstein Registry No.: 2691656


Purity: 97% CAS No.: 2905-24-0 Molecular Formula: C6H4BrClO2S Formula Weight: 255.52 Flash Point: 110°(230°F) Refractive Index: 1.5930 Melting Point: 30-33° Boiling Point: 90-91°/0.5mm Beilstein Reference: 2691656 UN No.: 3261 MDL No.: MFCD00052313

Formula: C₆H₄BrClO₂S
MW: 255.52 g/mol
Boiling Pt: 90…91 °C/0.5 mm
Melting Pt: 30…33 °C
Density: 1.773 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00052313
CAS Number: 2905-24-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR