6-Fluoroquinaldine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11469-06
A11469-22
A11469-14
AAA11469-06EA
72.8
USD
AAA11469-06
AAA11469-22
AAA11469-14
6-Fluoroquinaldine 98+%
6-Fluoroquinaldine
MDL: MFCD00041233
Beilstein Registry No.: 1281677
Purity: 98+%
CAS No.: 1128-61-6
Molecular Formula: C10H8FN
Formula Weight: 161.18
Melting Point: 52-56°
Beilstein Reference: 1281677
Formula:
C₁₀H₈FN MW: 161.18 g/mol Melting Pt: 52…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041233 CAS Number: 1128-61-6 EINECS: 214-439-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...