2,5-Dihydroxybenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Aspirin metabolite and antioxidant excipient
Purity: 99%
CAS No.: 490-79-9
Molecular Formula: C7H6O4
Formula Weight: 154.12
Melting Point: ca 206° dec.
Beilstein Reference: 2209119
Merck Reference: 14,4398
MDL No.: MFCD00002460
Formula:
C₇H₆O₄ Boiling Pt: 339 °C (1013 hPa) Melting Pt: ca 206 °C (dec.) Density: 1.55 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002460 CAS Number: 490-79-9 EINECS: 207-718-5 Merck Index: 13,04409 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...