1-Bromo-3-chloropropane 99%

Supplier: Thermo Scientific Chemicals
Danger

A11395-0B A11395-0I A11395-30 A11395-22
AAA11395-0BEA 92.1 USD
AAA11395-0B AAA11395-0I AAA11395-30 AAAA11395-22
1-Bromo-3-chloropropane 99%
1-Bromo-3-chloropropane

MDL: MFCD00000998 Beilstein Registry No.: 605278


Purity: 99% CAS No.: 109-70-6 Molecular Formula: Cl(CH2)3Br Formula Weight: 157.44 Flash Point: 140°(284°F) Refractive Index: 1.4855 Melting Point: -59° Boiling Point: 142-145° Beilstein Reference: 605278 UN No.: 2688 MDL No.: MFCD00000998

Formula: BrH₂CCH₂CH₂Cl
MW: 157.44 g/mol
Boiling Pt: 142…145 °C
Melting Pt: –59 °C
Density: 1.6 g/cm³ (20 °C)
Flash Pt: >140 °C (284 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000998
CAS Number: 109-70-6
EINECS: 203-697-1
UN: 2688
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR