D-(-)-Serine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (R)-(-)-Serine, H-D-Ser-OH

A11353-22 A11353-06 A11353-14
AAA11353-22EA 427 USD
AAA11353-22 AAA11353-06 AAA11353-14
D-(-)-Serine 99%
D-(-)-Serine

MDL: MFCD00004269 Beilstein Registry No.: 1721403 Fieser: 14,282 Optical Rotation: -13°C (c=5 in 5N HCl)


Purity: 99% CAS No.: 312-84-5 Molecular Formula: C3H7NO3 Formula Weight: 105.09 Melting Point: 218-220° dec. Beilstein Reference: 1721403 Merck Reference: 14,8460 Optical Rotation: -13° (c=5 in 5N HCl) MDL No.: MFCD00004269

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Melting Pt: 235 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004269
CAS Number: 312-84-5
EINECS: 206-229-4
Merck Index: 14,8460

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR