5-Chloro-2-thiophenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Chlorothiophene-2-sulphonyl chloride

A11348-09 A11348-06 A11348-03
AAAA11348-09EA 91.8 USD
AAAA11348-09 AAA11348-06 AAA11348-03
5-Chloro-2-thiophenesulfonyl chloride 97%
5-Chloro-2-thiophenesulphonyl chloride

MDL: MFCD00051667 Beilstein Registry No.: 130710


Purity: 97% CAS No.: 2766-74-7 Molecular Formula: C4H2Cl2O2S2 Formula Weight: 217.09 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5860 Boiling Point: 117-120°/14mm Beilstein Reference: 130710 UN No.: 3265 MDL No.: MFCD00051667

Formula: C₄H₂Cl₂O₂S₂
MW: 217.1 g/mol
Boiling Pt: 117…120 °C/14 mm
Melting Pt: 28 °C
Density: 1.623 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00051667
CAS Number: 2766-74-7
EINECS: 700-248-6
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR