Pentadecan-15-olide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cyclopentadecanolide, Oxacyclohexadecan-2-one, Pentadecanolide, ω-Pentadecalactone, omega-Pentadecalactone, Oxa-2-cyclohexadecanone

A11341-14 A11341-22
AAA11341-14EA 50.9 USD
AAA11341-14 AAA11341-22
Pentadecan-15-olide 98%
Pentadecan-15-olide

MDL: MFCD00039667 Beilstein Registry No.: 143710


Purity: 98% CAS No.: 106-02-5 Molecular Formula: C15H28O2 Formula Weight: 240.39 Flash Point: 110°(230°F) Refractive Index: 1.4700 Melting Point: 33-37° Boiling Point: 137°/2mm Beilstein Reference: 143710 Merck Reference: 14,3910 MDL No.: MFCD00039667

Formula: C₁₅H₂₈O₂
MW: 240.39 g/mol
Boiling Pt: 137 °C/2 mm
Melting Pt: 33…37 °C
Density: 0.918
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039667
CAS Number: 106-02-5
EINECS: 203-354-6
UN: 3077
ADR: 9,III
Merck Index: 14,3910

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR